Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade

Agar, Bacteriological Grade

Agar, Bacteriological Grade
Certificates
Agar, Bacteriological Grade

3 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Melting Pt:90 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to cream to light yellow to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤15 % (1 g, 105°C)
  • Solubility:
    (1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
  • Gel strength:
    ≥700 g/cm² (1.5 %)
  • Gel point:
    32 to 38°C (1.5 %)
  • Ash:
    ≤5 %
  • pH:
    6.0 to 7.5 (10 % suspension)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 85,50 €
    85,50 €
    1 * 250 g
       85,50 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 273,00 €
    273,00 €
    1 * 1 kg
       273,00 €

  • Plastic drum
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 1 100,00 €
    1 100,00 €
    1 * 5 kg
       1 100,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.