ContactUsLinkComponent
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Toxic
Health
Flammable
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Certificates
undefined
Toxic
Health
Flammable
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.4 %