Palladium(II) acetate 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3375-31-3
:
Formula:C₄H₆O₄Pd
MDL Number:MFCD00012453
:224.51 g/mol
Melting Pt:205 °C (dec.)
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to brown Powder
- Assay:46.9 to 48.5 % (Pd)
- Total trace metal impurities:≤1000 ppm
Specification Test Results
Frequently Bought Together