Adenosine 5'-monophosphate sodium salt hydrate 99%
: Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate(x:1:x), Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl phosphate hydrate, 5'-Adenylic acid,sodium salt, hydrate (9CI)
:149022-20-8
:
:C₁₀H₁₄N₅Na₂O₈P
:MFCD00065023
:409.200 g/mol
:Freezer
- Environmentally Preferable:
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤20.00 %
- HPLC:≥98.5 %
- pH:4.50 to 6.50
- Specific optical rotation:-41° to -43° (20°C, 589 nm) (c=2 in water)
Frequently Bought Together