

Iron(III) nitrate nonahydrate 99.9995% (trace metals basis)
ACRO436810250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Off-white to violet to purple Crystals
- Titration Iodometric:13.5 to 14.2 % (Fe)
- Total trace metal impurities:≤5 ppm
:
:404 g/mol
:125 °C (1013 hPa)
:47 °C
:1.68 g/cm³
:1466
:5.1,III
:GHS07
:GHS03
:Ambient
:7782-61-8
:Fe(NO₃)₃·9H₂O
:MFCD00149708