n-Hexadecane, anhydrous 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:544-76-3
:
Formula:H₃C(CH₂)₁₄CH₃
MDL Number:MFCD00008998
:226.45 g/mol
Boiling Pt:287 °C (760 mmHg)
Melting Pt:18 °C
Density:0.773 g/cm³
Flash Pt:135 °C
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- Water:≤0.005 %
- GC:≥98.5 %
- Color scale:≤10 APHA
Specification Test Results
Related Information
Frequently Bought Together