

Trifluoroacetic acid 99+% for spectroscopy
Catalog # ACRO434161000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- UV:at 320 nm A: ≤0.03
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Titration with NaOH:≥99.0 %
Specification Test Results
About this item
:
:114.02 g/mol
Boiling Pt:72 °C (760 mmHg)
Melting Pt:–15 °C
Density:1.535 g/cm³
UN:2699
ADR:8,I
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169