Order Entry
Belgium
ContactUsLinkComponent
4-Methyl-2-pentanone 99.5% for HPLC
Irritant
Flammable
4-Methyl-2-pentanone 99.5% for HPLC
Catalog # ACRO433570010
   Safer Human and Environmental Health
CAS Number:  
undefined
Irritant
Flammable
4-Methyl-2-pentanone 99.5% for HPLC
Catalog # ACRO433570010
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.4 %
  • UV:
    at 400 nm A: ≤0.01
  • Color scale:
    ≤15 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g

Specification Test Results

About this item

CAS Number:108-10-1
 :
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
 :100.16 g/mol
Boiling Pt:117.4 °C (760 mmHg)
Melting Pt:–84 °C
Density:0.8 g/cm³
Flash Pt:14 °C
Storage Temperature:Ambient
UN:1245
ADR:3,II
 :GHS07
 :GHS02