Order Entry
Belgium
ContactUsLinkComponent
 
Water ACS for spectroscopy
  ACRO433380010
   Safer Human and Environmental Health
 :  
undefined
Water ACS for spectroscopy
  ACRO433380010
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

 

 

  • Appearance:
    Clear colorless Liquid
  • UV:
    at 300 nm A: ≤0.005
  • Gradient analysis:
    Passes test
  • Absorbance:
    Passes test
  • KMnO4-reducing substance:
    passes test
  • Conductivity:
    2.0 x 10-6 ohm-1 cm-1

 

 

 :
 :18.02 g/mol
 :100 °C (760 mmHg)
 :0 °C
 :1 g/cm³
 :Ambient
 :7732-18-5
 :H₂O
 :MFCD00011332