Order Entry
Belgium
ContactUsLinkComponent
Trifluoroacetic acid 99+% for HPLC
Irritant
Corrosive
Trifluoroacetic acid 99+% for HPLC
Certificates
undefined
Irritant
Corrosive
Trifluoroacetic acid 99+% for HPLC
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • UV:
    at 260 nm A: ≤1.0
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤5 ppm
  • Titration with NaOH:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together