Order Entry
Belgium
ContactUsLinkComponent
4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane 97%
Corrosive
Irritant
4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane 97%
Catalog # ACRO431210010
CAS Number:  
undefined
Corrosive
Irritant
4,4,5,5-Tetramethyl-2-(1-methylpiperid-4-yl)-1,3,2-dioxaborolane 97%
Catalog # ACRO431210010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Yellow Low melting solid or liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • HPLC:
    ≥97.0 %

Specification Test Results

About this item

 :
Melting Pt:25…27 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
 :225.139 g/mol
 :GHS05
 :GHS07
CAS Number:1264198-72-2
Formula:C₁₂H₂₄BNO₂
MDL Number:MFCD11506064