ContactUsLinkComponent
1-Butanol, extra dry 99+%, AcroSeal®
Irritant
Corrosive
Flammable
1-Butanol, extra dry 99+%, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Flammable
1-Butanol, extra dry 99+%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.001 %