Ethanol absolute, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78 °C (760 mmHg)
Melting Pt:–114 °C
Density:0.79 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:clear colorless liquid
- Infrared spectrum:Conforms
- GC:≥99.5%
- Water:≤0.005%
- Residue after evaporation:≤0.001%
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO39769
SDS
ACRO397691000_TDS_EN
Product specification
Frequently Bought Together