Sodium-1-decanesulphonate, anhydrous 99+% for ion pair chromatography
:13419-61-9
:
:C₁₀H₂₁NaO₃S
:MFCD00007526
:244.33 g/mol
:>300 °C
:Ambient
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (150°C)
- UV:(0.2 M in water, 40°C)
- Titration after Ion exchange:≥99.0 % (on dry substance)
- Membrane filtration test:(on 0.2 M soln.) Passes test
- Suitability:(HPLC gradient) Passes test
Frequently Bought Together