
Ammonium hydrogen carbonate 99% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Melting Pt:105 °C (dec.)
Density:1,57 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Sulfated ash:≤0.05 %
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤5 ppm
- Phosphate (PO4):≤5 ppm
- Calcium (Ca):≤10 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤5 ppm
- Potassium (K):≤10 ppm
- Sodium (Na):≤20 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
Specification Test Results
Frequently Bought Together