
Tetrabutylammonium hydrogen sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:32503-27-8
:
Formula:(C₄H₉)₄NHSO₄
MDL Number:MFCD00011637
:339.54 g/mol
Melting Pt:168…172 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:169 to 173 °C
- Solubility:(10 % in water) Almost clear colorless
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together