Order Entry
Belgium
ContactUsLinkComponent
1-Propanol for HPLC
1-Propanol for HPLC
Certificates
undefined
1-Propanol for HPLC

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.9 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Non-volatile matter:
    ≤0.0003 %
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g

Specification Test Results

Related Information

Frequently Bought Together