Water for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Boiling Pt:100 °C (760 mmHg)
Melting Pt:0 °C
Density:1 g/cm³
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- UV:at 200 nm A: ≤0.05
- Residue after evaporation:≤0.0003 %
- Conductivity:≤1 µS/cm
Specification Test Results
Frequently Bought Together