Order Entry
Belgium
ContactUsLinkComponent
 
Hydrochloric acid ∼32% in water, pure
Catalog # ACRO389310025
CAS Number:  
undefined
Hydrochloric acid ∼32% in water, pure
Catalog # ACRO389310025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Residue after ign. (sulfated):
    ≤0.005 %
  • Titration with NaOH:
    ≥32 %
  • Sulfite (SO3):
    ≤5 ppm
  • Sulfate (SO4):
    ≤5 ppm
  • Arsenic (As):
    ≤0.5 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

About this item

 :
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.16 g/cm³
Storage Temperature:Ambient
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324