
Methyl decanoate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-42-9
:
Formula:C₁₁H₂₂O₂
MDL Number:MFCD00009580
:186.29 g/mol
Boiling Pt:226 °C (760 mmHg)
Melting Pt:–18 °C
Density:0.871 g/cm³
Flash Pt:90 °C
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4240 to 1.4270 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together