Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use

Buffer solutions, ready to use

Buffer solutions, ready to use
Certificates

6 Options Available Below

Product Details & Documents
  • Traceable to NIST
  • Clear colourless solutions
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ACRO383835000
  • ACRO383830010
  • ACRO383855000
  • ACRO383850010
  • ACRO383875000
  • ACRO383870010
  • Density:
  • 1
  • 1
  • 1
  • 1

Specifications

Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Colour
    Pack type
    Pk
    Availability
    Price
  • 4,00
    ±0,02
    Phthalate
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 38,40 €
    38,40 €
    1 * 500 mL
       38,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383835000
    Density
  • 4,00
    ±0,02
    Phthalate
    Colourless
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 61,30 €
    61,30 €
    1 * 1 L
       61,30 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383830010
    Density
  • 7,00
    ±0,02
    Phosphate
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 40,60 €
    40,60 €
    1 * 500 mL
       40,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383855000
    Density
    1
  • 7,00
    ±0,02
    Phosphate
    Colourless
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 74,20 €
    74,20 €
    1 * 1 L
       74,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383850010
    Density
    1
  • 9,2
    ±0,02
    Borate
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,10 €
    31,10 €
    1 * 500 mL
       31,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383875000
    Density
    1
  • 9,2
    ±0,02
    Borate
    Colourless
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 44,10 €
    44,10 €
    1 * 1 L
       44,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    ACRO383870010
    Density
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.