Order Entry
Belgium
ContactUsLinkComponent
 
 
Guanidinium hydrochloride 99.5%, without anti-caking agent
 
undefined
 
Guanidinium hydrochloride 99.5%, without anti-caking agent

 

  • Appearance:
    Colorless to white Crystals
  • Infrared spectrum:
    Conforms to structure and standard
  • Melting point:
    178 to 188 °C (Identification C)
  • UV:
    (6 M in water)
  • Identification A:
    Passes test
  • Identification B:
    Passes test
  • Titration with HClO4:
    ≥99.5 %

 

 

Frequently Bought Together