Order Entry
Belgium
ContactUsLinkComponent
Ammonium acetate 98+% for HPLC
Ammonium acetate 98+% for HPLC
Certificates
undefined
Ammonium acetate 98+% for HPLC

Specification Test Results

  • Appearance:
    White Powder or crystals
  • UV:
    (10 % in water)
  • pH:
    6.5 to 7.5 (5% soln.in water)
  • Titration with HClO4:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together