


Iodine 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Boiling Pt:185 °C (760 mmHg)
Melting Pt:113 °C
Density:4,9 g/cm³ (20 °C)
UN:3495
ADR:8,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark blue to black Glistening spheres
- Titration Iodometric:99.5 to 100.5 %
- Chlorine and Bromine (as Cl):≤0.005 %
- Calcium (Ca):≤10 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤5 ppm
- Magnesium (Mg):≤5 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤50 ppm
- Zinc (Zn):≤5 ppm
- Insoluble matter:Passes test
Specification Test Results
Frequently Bought Together