Sodium sulphate, anhydrous 99+% for HPLC
: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
:7757-82-6
:
:Na₂SO₄
:MFCD00003504
:>1700 °C (760 mmHg)
:884 °C
:2,7 g/cm³ (20 °C)
:142.04 g/mol
:Ambient
- Environmentally Preferable:
- Appearance:White Crystalline powder
- Water:≤0.1 %
- UV:at 220 nm A: ≤0.1
- Acidity/alkalinity:≤0.0002 meq/g
- Titration with HClO4:≥99.0 %
Frequently Bought Together