Sodium hydrogen sulphate monohydrate 99% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium bisulphate monohydrate
CAS Number:10034-88-5
:
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210
Molecular Weight:138.08 g/mol
Melting Pt:58 °C
Merck Index:13,08658
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Adhering crystals
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤0.0005 %
- Nitrate (NO3):≤0.001 %
- Phosphate (PO4):≤0.001 %
- Silicate (as SiO2):≤0.005 %
- Ammonium (NH4):≤0.001 %
- Iron (Fe):≤0.002 %
- Heavy metals (as Pb):≤5 ppm
- NH4OH precipitate:≤0.005 %
Specification Test Results
Frequently Bought Together