Colour standards (Pt-Co, Hazen) according to EN ISO 6271-1:2004
Supplier: Bernd Kraft
Certificates
About this item
ISO 6271-1:2004 specifies a method for estimating the colour, in Pt-Co units, of clear liquids.
Frequently Bought Together
ISO 6271-1:2004 specifies a method for estimating the colour, in Pt-Co units, of clear liquids.