VWR® RiboReady™ Color Micro RNA Ladder
N° de catalogue 1B1491-25RXN
Fournisseur: VWR Chemicals
Numéro CAS:
Spécifications
- Pk:1 KIT
- Description:RIBOREADY™ Color Micro RNA ladder
- Seq:01
Spécifications
À propos de cet article
RiboReady™ Color Micro RNA ladder allows precise sizing of RNA molecules in denaturing acrylamide gels. The apparent sizes of bands in the RiboReady™ Color Micro RNA ladder are 100, 75, 50, 40, 30 and 20 bases in length.
- 6 bands ranging from 20 to 100 bases
- Contains pre-stained nucleic acids that are visible during electrophoresis
Ready to use mixture that does not require heating or addition of a denaturing agent before use.