Haematoxylin solution (Mayer), Clin-Tech
Supplier: KLINIPATH
Certificates
About this item
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111
:302.28 g/mol
Related Information
Certificates
Search by Lot Numberspec_VWRK4085-9001 spec_VWRK4085-9002 spec_VWRK4085-9005 spec_VWRK4085.9001 spec_VWRK4085.9002
Spécification du produit
Plus
Frequently Bought Together