Orders LinkContactUsLinkComponent
Microscope slides for printers
Microscope slides for printers
Supplier:   KLINIPATH
Microscope slides for printers
Supplier:  KLINIPATH
Certificates
Microscope slides for printers
7 Options Available Below
Product Details & Documents
Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.
  • Frosted ends
  • 90° grounded edges
  • Finished clipped corners of 1mm
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 631-1229
    Microscope slides
    Microscope slides
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 65,80 €
    65,80 €
    1 * 2.500 ST
       65,80 €
  • 631-1231
    Microscope slides with frosted ends on both sides
    Microscope slides with frosted ends on both sides
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 76,10 €
    76,10 €
    1 * 2.500 ST
       76,10 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 207,00 €
    207,00 €
    1 * 2.500 ST
       207,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 207,00 €
    207,00 €
    1 * 2.500 ST
       207,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 134,00 €
    134,00 €
    1 * 2.500 ST
       134,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 134,00 €
    134,00 €
    1 * 2.500 ST
       134,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 134,00 €
    134,00 €
    1 * 2.500 ST
       134,00 €