Orders LinkContactUsLinkComponent
pH electrodes, Eutech
pH electrodes, Eutech
Supplier:   EUTECH
pH electrodes, Eutech
Supplier:  EUTECH
Certificates
pH electrodes, Eutech
pH electrodes, Eutech
8 Options Available Below
Product Details & Documents
pH electrodes for use with all pH meters with BNC input connector.
Product Family Options
Product Information
  • pH
    Temp.
    Material
    Diaphr.
    Ø×L
    Connection
    For
    Availability
    Price
  • EUTEECFC7252101B
    pH electrode, single junction, sealed
    pH electrode, single junction, sealed
    1 - 13
    0...+80 °C
    Epoxy
    Porous HDPE pin
    12×90 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,11 €
    99,11 €
    1 * 1 ST
       99,11 €
  • EUTEECFC7252201B
    pH electrode, double junction, sealed
    pH electrode, double junction, sealed
    1 - 13
    0...+80 °C
    Epoxy
    Porous HDPE pin
    12×90 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 133,10 €
    133,10 €
    1 * 1 ST
       133,10 €
  • EUTEECFE7372801W
    pH combination electrode, double junction, sealed, built-in ATC
    pH combination electrode, double junction, sealed, built-in ATC
    1 - 13
    0...+80 °C
    Epoxy
    Porous HDPE pin
    12×115 mm
    BNC, 1 m cable
    General purposes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,80 €
    184,80 €
    1 * 1 ST
       184,80 €
  • EUTEECFG7350401B
    pH combination electrode, single junction, refillable, with protective sensor guard
    pH combination electrode, single junction, refillable, with protective sensor guard
    0 - 13
    0...+100 °C
    Glass
    Annular ceramic
    12×130 mm
    BNC, 1 m cable
    General purposes (samples with low temperature measurements)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,10 €
    144,10 €
    1 * 1 ST
       144,10 €
  • EUTEECFG7370101B
    pH combination electrode, double junction, refillable, clog-free
    pH combination electrode, double junction, refillable, clog-free
    0 - 13
    0...+100 °C
    Glass
    Annular ceramic
    12×110 mm
    BNC, 1 m cable
    Samples containing TRIS buffers, sulfides and more
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 297,00 €
    297,00 €
    1 * 1 ST
       297,00 €
  • EUTEECFG7451901B
    pH electrode, rugged, single junction, sealed
    pH electrode, rugged, single junction, sealed
    0 - 14
    5...+110 °C
    Glass
    Annular ceramic
    12×110 mm
    BNC, 1 m cable
    Strong alkaline solutions, photographic chemicals
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 289,30 €
    289,30 €
    1 * 1 ST
       289,30 €
  • EUTEECFG6351101B
    pH combination electrode, single junction, sealed
    pH combination electrode, single junction, sealed
    0 - 13
    0...+100 °C
    Glass
    Annular ceramic
    8×55 mm
    BNC, 1 m cable
    Solid / semi-solid samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 289,30 €
    289,30 €
    1 * 1 ST
       289,30 €
  • EUTEECFG6350601B
    pH electrode, single junction, refillable (with solution ECRE001)
    pH electrode, single junction, refillable (with solution ECRE001)
    0 - 13
    0...+100 °C
    Glass
    Annular ceramic
    8×55 mm
    BNC, 1 m cable
    High viscosity samples where frequent cleaning of reference is required
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,90 €
    240,90 €
    1 * 1 ST
       240,90 €