ContactUsLinkComponent
Storage tubes, 2D barcoded
Storage tubes, 2D barcoded
Catalog # MATRAB-1470
   Low Manufacturing Impact
Storage tubes, 2D barcoded
Catalog # MATRAB-1470
   Low Manufacturing Impact
Specifications
  • Cap included:
    Yes
  • Capacity:
    5,00 ml
  • Description:
    2D barcoded tubes, with caps, racked
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    240
Specifications
About this item
Unique and laser etched 2D barcode is encapsulated on the base to securely identify and track samples.
  • Grid-referenced lids enable storage tube location
  • External threads tubes aid in contamination prevention
  • Save space with dense microplate footprint
The laser-etched 2D barcode feature scan the tube into any application or database, or integrate via various data export options. Every storage tube code is scanned to guarantee readability. Each code is checked against the entire database of all 2D barcoded storage tubes to ensure zero duplicates. Every lot of storage tubes is leak tested to ensure the integrity and security of samples. All tubes are certified free of DNase, RNase and DNA.