Order Entry
Belgium
Orders LinkContactUsLinkComponent
Soil sampler

Soil sampler

Soil samplers, Plastic, Natural, 25 mm
Catalog # EPSCTERRACORE5G
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Soil sampler
Catalog # EPSCTERRACORE5G
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Soil sampler
Soil sampler

Soil samplers, Plastic, Natural, 25 mm

Price
931,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

This rugged all-in-one item eliminates the need for a separate handle.

  • More convenient than make-your-own cut-off syringe
  • Simple operation for easy and effective sampling
  • Minimizes handling and reduces target analyte loss

Designed for soil sampling into a 40 ml VOA vial for in-field preservation.

Specifications
  • Colour:
    Natural
  • Description:
    Soil samplers
  • Length:
    25 mm
  • Material:
    Plastic
  • Cat. No.:
    EPSCTERRACORE5G
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.