Orders LinkContactUsLinkComponent
Dust covers
Dust covers
Dust covers, for VOA vial caps, 24 - 414 mm
Catalog # EPSCDC-VOA
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Dust covers
Catalog # EPSCDC-VOA
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Dust covers
Dust covers
Dust covers, for VOA vial caps, 24 - 414 mm
Price
220,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
These PE dust covers are designed to fit all standard size VOA vial caps. They prevent contamination and protect critical samples during transport.
  • Allow for septum expansion
  • Pre cleaned dust covers available upon request
Specifications
  • Cap Size:
    24 - 414 mm
  • Description:
    Dust covers, for VOA vial caps
  • Cat. No.:
    EPSCDC-VOA
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact