Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Centrifuges
Rotors
Rotor for centrifugal evaporators, RC10-10 and RC10-22, Jouan
Rotor for centrifugal evaporators, RC10-10 and RC10-22, Jouan
Rotor for centrifugal evaporators, RC10-10 and RC10-22, Jouan
Catalog # 531-0025
Supplier:
JOUAN
Expand image
Rotor for centrifugal evaporators, RC10-10 and RC10-22, Jouan
Catalog # 531-0025
Supplier:
JOUAN
Specifications
Description:
Rotor, sandwich, for 10×50 ml tubes (max. 29×103 mm)
Pk:
1
Specifications
About this item
Product Details
Product Details