Orders LinkContactUsLinkComponent
Heated forceps, eTemp
Heated forceps, eTemp
Supplier:   KLINIPATH
Heated forceps, eTemp
Supplier:  KLINIPATH
Certificates
Heated forceps, eTemp
5 Options Available Below
Product Details & Documents
eTemp is an electrically heated forceps system that can be used in the preparation of histological or histopathological samples.
  • Housing made from welded stainless steel and powder coated
  • No temperature adjustment
  • No tiresome reheating — full attention on processing samples
  • Faster embedding increases productivity
  • No contact problems at forceps connection
To ensure that small amounts of preparations do not remain on cooled forcep tips, the forceps must be repeatedly removed from the preparation and heated via external means such as a flame or an embedding station. The eTemp operates at a safe low voltage and features temperature-regulated heating elements that constantly keep the forcep tips at the optimum temperature for handling preparations in histological paraffin. This model features all-metal forceps with medical plastic insulation, which are heated exclusively at the tips.
Product Family Options
Forceps
Product Information
  • Availability
    Price
  • KLIN4509205
    Straight forceps, 0.5 mm
    Straight forceps, 0.5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • KLIN4509210
    Straight forceps, 1.0 mm
    Straight forceps, 1.0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • KLIN4509210M
    Straight forceps, 1.0 mm, MCB
    Straight forceps, 1.0 mm, MCB
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • KLIN4509220
    Straight forceps, 2.0 mm
    Straight forceps, 2.0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • KLIN4509240
    Straight forceps, 4.0 mm
    Straight forceps, 4.0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €