ContactUsLinkComponent
Visor for helmet attachment, uvex 9790
Visor for helmet attachment, uvex 9790
Catalog # 111-1883
Supplier:  UVEX
undefined
Visor for helmet attachment, uvex 9790
Catalog # 111-1883
Supplier:  UVEX
Specifications
  • Description:
    Adapter for attaching visor to helmet without helmet earmuffs
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Helmet visor made from strong PC protects against the effects of electrical arcs. It is compatible with uvex safety helmets and uvex 2H, uvex 3H and uvex 3200H earmuffs with helmet attachment.
Ordering information: The visor is supplied without helmet, earmuffs and bracket. All parts can be ordered separately. Please note: An additional adapter (UVEX9924.010) is required for helmet and visor combination without helmet earmuffs.
Suitable earmuffs
Product Information