Orders LinkContactUsLinkComponent
Labware value pack, Nalgene®
Labware value pack, Nalgene®
Labware value pack
Catalog # 212-0440
   Low Manufacturing Impact
Labware value pack, Nalgene®
Catalog # 212-0440
   Low Manufacturing Impact
Labware value pack, Nalgene®
Labware value pack, Nalgene®
Labware value pack
Price
187,00 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Labware Value Pack is a useful assortment of popular labware items used in every lab.
  • Serves as new lab starter or refresher pack
  • Sets the standard for quality and superior performance with the highest quality materials
  • Shatterproof and it provides a safer, longer equipment life at lower replacement costs than glass
Note: Remove jar closure and autoclave separately. LDPE Unitary™ wash bottles are not autoclavable. All other items in Value Pack are autoclavable.
Certifications: ASTM E1272, CFR 21, ISO 6706, Part 177.1520.
Ordering information: Includes six straight-sided jars (500 ml/PPCO), four Unitary™ wash bottles (500 ml/LDPE), four graduated cylinders (2 to 100 ml, 500 ml, 1 L/PPCO), five Griffin low form beakers (50, 100, 250, 600, 1000 ml/PPCO), Unwire™ blue half-rack for 30 mm tubes.
Spécifications
  • Description:
    Labware value pack
  • Cat. No.:
    212-0440
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact