Order Entry
Belgium
ContactUsLinkComponent
 
Purification systems, KingFisher™ Flex
  LABL5400630
Purification systems, KingFisher™ Flex
  LABL5400630

 

  • Plate types:
    96 deep well plate (50 - 1000 µl), KingFisher 96 plate
    (20 - 250 µl), PCR plate (20 - 100 µl)
  • Description:
    Purification system KingFisher™ Flex with 96 deep well head
  • Model:
    KingFisher™ flex
  • Interfaces:
    USB or serial RS232C port
  • Int. software:
    Space for ~ 100 internal protocols
  • Keyboard/display:
    START/STOP/PAUSE/OK, four arrow keys,
    two keys to rotate the turntable
  • Magnet rods:
    96
  • Particle collection efficiency:
    ≥95%
  • Processing volume:
    20 - 1000 µl
  • Robot compatibility:
    Yes
  • Tip combs (disposable):
    Special design, 96 format
  • Weight:
    28 kg
  • W×D×H:
    680×600×380 mm
  • Capacity (samples/run):
    96
  • Magnetic particle size:
    ~ >1 μm
  • Pk:
    1

 

 

The KingFisher™ magnetic particle processors are designed to automate time consuming sample processing of nucleic acids, proteins or cells from virtually any source (blood, cell cultures, tissue lysates, soil, faeces, for example). The KingFisher™ family consists of instruments with different throughput and volume capacities to fulfil individual sample processing needs. Each system consists of an instrument, specially designed plastics, and software. The KingFisher™ systems offer rapid and reproducible purification of high quality DNA, RNA, proteins and cells for various types of downstream applications, including genomics and proteomics, drug discovery, forensics, biomarker discovery, quality control, and veterinary assays.

  • High speed, fully automated purification
  • Purification of any sample from any source
  • Ready made and custom protocols
  • Easy operation via instrument or external PC

KingFisher™ Flex offers highly versatile, automated magnetic particle processing for DNA/RNA, protein or cell purification from virtually any source. The system is able to handle a range of volumes from 20 to 5000 μl, depending on the magnet head, using either a 96- or 24-well plate format. Supplied with predefined protocols, KingFisher Flex allows the creation of custom-made protocols for special applications.