Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flow cytometry fixation/permeabilisation kit

Flow cytometry fixation/permeabilisation kit

Flow Cytometry Fixation/Permeabilisation Kit, 50 Tests
Catalog # 23006.
Supplier:   Biotium
Flow cytometry fixation/permeabilisation kit
Catalog # 23006.
Supplier:  Biotium
CAS Number:  
Flow cytometry fixation/permeabilisation kit
Flow cytometry fixation/permeabilisation kit

Flow Cytometry Fixation/Permeabilisation Kit, 50 Tests

CAS Number:  
Price
49,90 €
Log in for availability
1 * 50 Tests
Product Details & Documents

Flow Cytometry Fixation/Permeabilisation Kit contains optimally formulated buffers for fixation and permeabilisation of suspension cells for immunofluorescence staining of intracellular antigens for analysis by flow cytometry. The bufffers are also available separately.

Delivery information: Kit contains Fixation Buffer, 5 ml and Permeabilisation Buffer, 5 ml.

Documents

Specifications

Specifications

  • Pk:
    50 Tests
  • Description:
    Flow Cytometry Fixation/Permeabilisation Kit

Specifications

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Fixation buffer, ready to use
    Fixation buffer, ready to use
    22015.
    100 mL
    Log in for availability
    1 * 100 mL - 173,00 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Cell permeabilisation buffer, ready-to-use
    Cell permeabilisation buffer, ready-to-use
    22016.
    100 mL
    Log in for availability
    1 * 100 mL - 173,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.