Pipette tips, Matrix™ SerialMate™
Supplier: THERMO MATRIX TECHNOLOGIES
Pipette tips, Matrix™ SerialMate™
Supplier: THERMO MATRIX TECHNOLOGIES
Certificates
About this item
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
7 Options Available Below
Product Details & Documents
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
Specifications for Product Family
Specifications
- Catalog No:
- MATR5541
- MATR5542
- MATR5545
- MATR5561
- MATR5562
- MATR5571
- MATR5572
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
- Tip styleVolumeSterileFilterLengthFor use withPackedAvailabilityPrice
- Standard5,0 - 220 µl--45,72 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST237,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5541Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Standard5,0 - 220 µl+-45,72 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST261,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5542Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Filtered5,0 - 220 µl++45,72 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST415,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5545Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Deep-well tips (extended)5,0 - 220 µl--57,91 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST237,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5561Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Deep-well tips (extended)5,0 - 220 µl+-57,91 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST262,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5562Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Standard0,5 - 30 µl--48,26 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST270,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5571Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - Standard0,5 - 30 µl+-48,26 mmMatrix™ SerialMate™20 racks, 96 eachLog in for availability1 * 1.920 ST299,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.MATR5572Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact