Order Entry
Belgium
ContactUsLinkComponent
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Irritant
Flammable
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
   Sustainable Options Available
Certificates
undefined
Irritant
Flammable
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.2 % (K.F.)
  • Boiling point:
    81 to 83 °C
  • Density:
    (20/20 °C) 0.784 to 0.787
  • Solubility:
    (in water) Passes test
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Carbonyl compounds:
    ≤0.002 % (propionaldehyde)
  • Titrable acid or base:
    ≤0.0001 meq/g

Specification Test Results

Related Information

Frequently Bought Together