



Sodium methoxide 30% (w/w) 5.4 M in methanol, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Melting Pt:1…5 °C
Density:0.97 g/cm³
Flash Pt:33 °C
UN:1289
ADR:3,III
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to light yellow Solution
- Strength:5.3 to 5.6 M
- HPLC:≥97.5 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO42836
SDS
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together