Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether 99.8% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Irritant
Flammable
tert-Butyl methyl ether 99.8% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Catalog # ACRO428010025
CAS Number:  
undefined
Irritant
Flammable
tert-Butyl methyl ether 99.8% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Catalog # ACRO428010025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • Density:
    (20/4°C) 0.740 to 0.742
  • GC:
    ≥99.75 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %
  • Peroxides:
    ≤1 ppm (as H2O2)

Specification Test Results

About this item

CAS Number:1634-04-4
 :
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812
 :88.15 g/mol
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–110 °C
Density:0.7404 g/cm³
Flash Pt:–28 °C
Storage Temperature:Ambient
UN:2398
ADR:3,II
 :GHS07
 :GHS02