Order Entry
Belgium
ContactUsLinkComponent
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Certificates
undefined
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (Coulometric)
  • Boiling point:
    109 to 111 °C
  • Density:
    (20/20 °C) 0.865 to 0.870
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Substance darkened by H2SO4:
    passes test
  • Sulfur compounds (as S):
    ≤0.003 %

Specification Test Results

Related Information

Frequently Bought Together