ω-Bromo-2-nitroacetophenone 98%
Glass bottle, 5 g
Fournisseur: Thermo Fisher Scientific
ω-Bromo-2-nitroacetophenone 98%
N° de catalogue ACRO398850050
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 6851-99-6
Jump to:
Product Details & Documents
Synonyms: α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide
- CAS Number:6851-99-6
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00010294
- Molecular Weight:244.04 g/mol
- Melting Pt:55…57 °C
- Storage Temperature:Ambient
- UN:3261
- ADR:8,II
Documents
ACRO39885
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:53 to 58 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



