Order Entry
Belgium
ContactUsLinkComponent
1-Propanol, extra dry 99.5%, AcroSeal®
1-Propanol, extra dry 99.5%, AcroSeal®
Certificates
undefined
1-Propanol, extra dry 99.5%, AcroSeal®

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.5 %
  • Residue after evaporation:
    ≤0.001 %
  • Filtered to 0.2 micrometer:
    Filtered

Specification Test Results

Frequently Bought Together