Hydrochloric acid 1 M in ethyl acetate
Catalog # ACRO388701000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Strength:0.95 to 1.15 M
Specification Test Results
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:0.9 g/cm³
Flash Pt:–3 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:36.46 g/mol
:GHS05
:GHS02
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324