Order Entry
Belgium
ContactUsLinkComponent
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
   Sustainable Options Available
Certificates
undefined
1-Butanol 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 % (K.F.)
  • Boiling point:
    116 to 119 °C
  • Density:
    (20/20 °C) 0.809 to 0.812
  • GC:
    ≥99.4 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
  • Carbonyl compounds:
    ≤0.01 % (as butyraldehyde)
  • Titrable acid:
    ≤0.0008 meq/g
  • Butyl ether:
    ≤0.2 % (GC)

Specification Test Results

Related Information

Frequently Bought Together