beta-Alanyl-L-histidine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 305-84-0
- Infrared spectrum: Conforms
- Water: ≤1%
- Specific optical rotation: 20,0 to 22,0° (20 °C, 589 nm) (c=2, water)
- Melting point: 253 °C (dec.)
Specifications
- CAS No.:305-84-0
- Storage conditions:Refrigerator
- Form:White to off-white powder
- MDL No.:MFCD00005207
- Protein synonyms:L-Ignotine|L-Carnosine|beta-Alanyl-L-histidine
- Protein/peptide name:β-Alanyl-L-histidine
- Purity:≥97,5% (Titration with HClO₄)
- Molecular weight:226,24 g/mol
- Molecular formula:C₉H₁₄N₄O₃
Specifications
Frequently Bought Together